Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OVOL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552037
Description
Immunogen sequence: RAFLVKKPCV STCKRNWSEL PDEERGEIYV PVSLGFCPPQ PYREPEPSVA EPPSCPLALN MSLRDSSYSM APGPCVVAQL PSEDMGHLTD PQSRDHGFLR TK Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 89%.
OVOL1 is a protein highly similar to Drosophila and mouse proteins. In Drosophila the ovo protein plays a critical role in Drosophila oogenesis and cuticle formation. In mice the ovo like protein is involved in hair formation and spermatogenesis. The function of the human gene product has not been determined.This gene encodes a protein highly similar to Drosophila and mouse proteins. In Drosophila the ovo protein plays a critical role in Drosophila oogenesis and cuticle formation. In mice the ovo like protein is involved in hair formation and spermatogenesis. The function of the human gene product has not been determined.
Specifications
OVOL1 | |
Polyclonal | |
Unconjugated | |
OVOL1 | |
BB147136; CD254; hOvo1; hRANKL2; Ly109l; movo1; mOvo1a; ODF; OPG; OPG ligand; OPGL; OPTB2; osteoclast differentiation factor; Osteoprotegerin ligand; OVO homolog-like 1; OVO homolog-like 1 (Drosophila); ovo like transcriptional repressor 1; ovo like zinc finger 1; Ovo1; Ovol1; OVO-like 1 binding protein; ovo-like 1(Drosophila); ovo-like zinc finger 1; Putative transcription factor Ovo-like 1; RANKL; receptor activator of NF-kappaB ligand; receptor activator of nuclear factor kappa B ligand; receptor activator of nuclear factor kappa-B ligand; sOdf; TNF-related activation-induced cytokine; TNFSF11; TNLG6B; Trance; transcription factor Ovo-like 2; tumor necrosis factor (ligand) superfamily member 11; tumor necrosis factor (ligand) superfamily, member 11; tumor necrosis factor ligand 6B; tumor necrosis factor ligand superfamily member 11; Tumor necrosis factor ligand superfamily member 11, membrane form; Tumor necrosis factor ligand superfamily member 11, soluble form; tumor necrosis factor superfamily member 11; tumor necrosis factor-related activation-induced cytokine | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5017 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O14753 | |
OVOL1 | |
Recombinant protein corresponding to Human OVOL1. Recombinant protein control fragment (Product #RP-102303). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction