Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OVOL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541619
Description
Peptide sequence: PKVFLVKRRS LGVSVRSWDE LPDEKRADTY IPVGLGRLLH DPPEDCRSDG Sequence homology: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 93%.
DNA-binding protein that binds to the 5'-G[GCT]GGGGG-3' core sequence. Probably acts as a transcription regulator.
Specifications
| OVOL2 | |
| Polyclonal | |
| Unconjugated | |
| OVOL2 | |
| 1700108N11Rik; 1810007D21Rik; EUROIMAGE566589; hOvo2; MOVO; M-OVO; movo2; M-OVO-A; M-OVO-B; MOVO-B zinc finger protein; MOVO-C zinc finger protein; OVO homolog-like 2; ovo like zinc finger 2; Ovo2; OVOL2; ovo-like 2; ovo-like 2 (Drosophila); ovo-like zinc finger 2; PPCD1; transcription factor Ovo-like 2; Zfp339; zinc finger OVO2; Zinc finger protein 339; zinc finger protein mOVO; Znf339 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 58495 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9BRP0 | |
| OVOL2 | |
| Synthetic peptide directed towards the N-terminal of human OVOL2 (aa 2-51). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction