Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OVOL2/ZNF339 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982095
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2095 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2095 Supplier Bioss Supplier No. BS12274R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DNA-binding protein that binds to the 5′-G[GCT]GGGGG-3′ core sequence. Probably acts as a transcription regulator.

Specifications

Antigen OVOL2/ZNF339
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene OVOL2
Gene Accession No. Q9BRP0
Gene Alias 1700108N11Rik; 1810007D21Rik; BOS_12956; EUROIMAGE566589; hOvo2; MOVO; M-OVO; mOvo2; M-OVO-A; M-OVO-B; MOVO-B zinc finger protein; MOVO-C zinc finger protein; OVO homolog-like 2; ovo like zinc finger 2; Ovo2; Ovol2; ovo-like 2; ovo-like 2 (Drosophila); ovo-like zinc finger 2; PPCD1; Transcription factor Ovo-like 2; Zfp339; Zinc finger OVO2; Zinc finger protein 339; Zinc finger protein mOVO; Znf339
Gene Symbols OVOL2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human OVOL2/ZNF339.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58495
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.