Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OVOL3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144818
Description
Immunogen sequence: CSLEAHLAKV HGQPASYAYR ERREKLHVCE DCGFTSSRPD TYAQH Antigen sequence identity: Mouse 98%; Rat 96%.
OVOL3 is a transcription factor that plays a crucial role in the development and differentiation of various tissues, including the skin, hair, and eyes. It is involved in regulating the expression of genes that control cell proliferation, differentiation, and apoptosis.
Specifications
OVOL3 | |
Polyclonal | |
Unconjugated | |
OVOL3 | |
HOVO; OVO3; OVO-like zinc finger 3; Zinc finger protein OVO-like 3 | |
Rabbit | |
Affinity chromatography | |
RUO | |
728361 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O00110 | |
OVOL3 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 163-207. Recombinant protein control fragment (Product #RP-110085). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction