Learn More
Owen Mumford™ Unistik™ 3 Gentle Single use Safety Lancets
Shop All Owen Mumford, Inc. Products
Click to view available options
Packaging Quantity:
100/Pk.
200/Pk.
25/Pk.
50/Pk.
Description
- Featuring proprietary Comfort Zone Technology™ for virtually pain-free capillary blood sampling
- Special design features side release button and preset lancet speed to ensure ease-of-use for healthcare professionals and maximum comfort for patients
- Simply twist off protective sterile cap, position and activate release button
- Needle point is hidden before use and automatically retracts after use to avoid needlestick injuries and cross infection
- Single use device designed to ensure highest level of safety for healthcare professionals and patients
Specifications
Specifications
| Needle Gauge | 30G |
| Packaging Quantity | 50/Pk. |