Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Owen Mumford™ Unistik™ 3 Single-Use Safety Lancets
Shop All Owen Mumford, Inc. Products

Click to view available options
Color:
Lavendar
Orange
Depth (Metric):
1.8mm
2mm
Needle Gauge:
21g
28g
Quantity:
100 Pack
1000 Case
200 Pack
Description
- Requires no cocking procedure – just twist off protective sterile cap and use
- Convex platform requires less technique and training for use
- Ergonomically designed with special indentions on each side for finger placement
- Needle point is hidden before use and automatically retracts after use to avoid accidental needlestick injuries and cross-infection
- Rendered inoperable after use, providing highest level of safety for healthcare workers and patients and meeting OSHA standards
- Visual lockout feature clearly indicates that device has been used and is ready for disposal in a sharps container
- High-flow volume suitable for cholesterol, hemoglobin and cardiac enzyme testing
US Patents 5,487,748; 6,258,112; 6,764,496
Specifications
Specifications
| Disposable | Single-use |
| For Use With (Application) | For ward blood glucose monitoring, HbA1c |
| Quantity | 200 Pack |
| Product Type | Unistik™ 3 Safety Lancet |
| Needle Gauge | 21g |
| Depth (Metric) | 2mm |
| Depth (English) | 0.07 in. |
| Color | Orange |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction