Learn More
Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System
Description
Run 10 to 80 samples on one gel with the Thermo Scientific™ Owl™ D2 Wide-Gel Electrophoresis System.
- Wide gel size allows 15 or 30 samples to be loaded per row with a micro well comb.
- Using special combs and a multichannel pipetter, samples may be loaded directly from a 96-well plate, eight or 12 at a time.
- System is excellent for screening PCR samples or routine DNA fragment analysis.
- Small footprint saves space on lab bench.
- Incorporates Thermo Scientific™ EasyCast™ casting design with external caster—gel tray is fitted with gasket, allowing it to fit snugly into caster and providing a leakproof seal without tape.
- Electrode placement allows gel to be run quickly without compromising resolution.
Includes:
- Buffer chamber
- Thermo Scientific™ SuperSafe™ lid with attached power supply leads
- EasyCast gasketed UVT gel tray
- External multiple gel caster
- Four combs (two 30-well and two 40-well, 1.5mm thick)
Specifications
Specifications
| Certifications/Compliance | CE marked |
| Description | D2 Wide-Gel |
| Product Line | Owl |
| Type | Wide |
| Sample Size | 10 to 80 |
| Volume (Metric) Buffer | 600 mL |
| Dimensions (L x W x H) | 7.38 x 7.75 x 4 in. (19 x 20 x 10 cm) |
| Length (English) | 7.38 in. |
| Length (Metric) | 19 cm |
| Length (Metric) Gel | 14.4 cm |
| Show More |
Frequently Asked Questions (FAQs)
The Owl Electrophoresis Systems are supplied with standard power leads, but we have not tested compatibility of the Owl systems with the Bio-Rad power supplies. For best performance, we recommend using one of our Owl or PowerEase power supplies for the Owl Electrophoresis Systems.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.