Learn More
Thermo Scientific™ Owl™ EasyCast™ B1 Mini Gel Electrophoresis Systems
Description
Cast and run a gel in the same chamber with no tape or additional parts required with the leakproof casting of Thermo Scientific™ Owl™ EasyCast™ B1 Mini Gel Electrophoresis Systems. Durability and ease of use makes this the perfect choice for students and high volume researchers.
Features:
- System offers a wide range of combs.
- Two comb slots on gel tray double sample capacity by doubling number of sample wells.
- UVT gel trays are silk screened with a florescent ruler for easy measurement of bands.
Includes:
- Main buffer chamber
- Thermo Scientific™ SuperSafe™ lid with attached power supply leads
- Thermo Scientific™ EasyCast™ Gasketed UVT gel tray
- Two Combs: 10 & 14 Well, Double-sided, 1.0/1.5mm thick
Ordering Information:
Available with optional buffer exchange ports, (B1-BP), for use with an external recirculation pump.
Warranty and Service Offering:
- Three years
Specifications
Specifications
| Certifications/Compliance | CE marked |
| Gel Size | Mini |
| Product Line | Owl |
| Type | Mini Gel Electrophoresis System |
| Includes | Without Buffer Exchange Ports |
| Sample Size | 5 to 34 |
| Volume (Metric) Buffer | 600 mL |
| Dimensions (L x W x H) | 8.63 x 5.88 x 3.75 in. (22 x 15 x 9.5 cm) |
| Length (English) | 8.63 in. |
| Length (Metric) | 22 cm |
| Show More |
Frequently Asked Questions (FAQs)
The Owl Electrophoresis Systems are supplied with standard power leads, but we have not tested compatibility of the Owl systems with the Bio-Rad power supplies. For best performance, we recommend using one of our Owl or PowerEase power supplies for the Owl Electrophoresis Systems.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.