Learn More
Thermo Scientific™ Owl™ EasyCast™ B1A Mini Gel Electrophoresis Systems
Description
Cast and run a gel in the buffer chamber and achieve flat, even banding patterns on a consistent basis with Thermo Scientific™ Owl™ EasyCast™ B1A Mini Gel Electrophoresis Systems. The ideal size for every lab, this system determines molecular weight ranges of PCR fragments or larger sized DNA molecules.
- Two comb slots on gel tray double sample capacity by doubling number of sample wells.
- UVT gel trays are silk screened with a florescent ruler for easy measurement of bands.
- Leakproof casting allows a gel to be cast and run in the same chamber with no tape or additional parts required
- Run from five to 24 samples in 15 minutes.
Ordering Information:
Available with optional buffer exchange ports (B1A-BP) for use with an external recirculation pump.
Includes:
- Main buffer chamber
- Thermo Scientific™ SuperSafe™ lid with attached power supply leads
- Thermo Scientific™ EasyCast gasketed UVT gel tray
- Two heavy-duty, one-piece, double-sided combs: 6 & 10 well, 1.0/1.5mm thick
Specifications
Specifications
| Certifications/Compliance | CE marked |
| Gel Size | Mini |
| Product Line | Owl |
| Type | Mini Gel Electrophoresis System |
| Includes | EasyCast |
| Sample Size | 5 to 24 |
| Volume (Metric) Buffer | 400 mL |
| Dimensions (L x W x H) | 6.31 x 4.13 x 3.75 in. (16 x 10.5 x 9.5 cm) |
| Length (English) | 6.31 in. |
| Length (Metric) | 16 cm |
| Show More |
Frequently Asked Questions (FAQs)
The Owl Electrophoresis Systems are supplied with standard power leads, but we have not tested compatibility of the Owl systems with the Bio-Rad power supplies. For best performance, we recommend using one of our Owl or PowerEase power supplies for the Owl Electrophoresis Systems.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.