Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Innovating Science™ Owl Pellet Kit

Shop All Aldon Products


Click to view available options
Class Size:
5 Lab Groups
Description
- Owls have a specialized digestive tract which helps them expel the indigestible parts of their prey in pellet form
- The pellets are heat sterilized and individually wrapped
- Kit Includes:
- 5 Owl Pellets
- 10 Wooden Probes
- 5 Forceps
- 5 Plastic Rulers
- 5 Hand Magnifiers
- 20 Nitrile Gloves
- Teacher's Manual and Guide
Specifications
Specifications
For Use With (Application) | Groups of 5 Students |
Size | Small |
Length (English) | 9.75 in. |
Class Size | 5 Lab Groups |
Product Type | Owl Pellet Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction