Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hand2Mind Owl Pellet Mystery Science Lab Kit (BONES)

Click to view available options
Class Size:
10 Students
Description
- Explore the world of biology and perform your own experiments
- Discover what it's like to be a wildlife biologist or veterinarian.
- Use the included lab tools to dissect real owl pellets and turn your home into your very own bio lab.
- STEM at Play™ kits bring science concepts to life through hands-on activities designed to spark interest, inspire confidence, and prompt creative critical-thinking skills while fostering collaboration between children and adults.
- STEM at play kits provide materials to support scholastic goals while keeping at-home learning fun.
Specifications
Specifications
Grade Level | Elementary, Intermediate - Middle School, High School |
Grade | K to 12 |
Class Size | 10 Students |
Product Type | Owl Pellet Mystery Lab Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction