Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OX40 Ligand/TNFSF4 Antibody (159403) [DyLight 680], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB10541Y
Description
OX40 Ligand/TNFSF4 Monoclonal antibody specifically detects OX40 Ligand/TNFSF4 in Human samples. It is validated for Flow Cytometry, Neutralization, Immunocytochemistry, CyTOFSpecifications
OX40 Ligand/TNFSF4 | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Neutralization, Immunocytochemistry, CyTOF | |
159403 | |
Flow Cytometry, Neutralization, Immunocytochemistry, CyTOF-ready | |
CD134 ligand, CD134L, CD252, CD252 antigen, Glycoprotein Gp34, gp34, OX40 antigen ligand, OX40 ligand, OX40L, OX-40L, OX4OL, TAX transcriptionally-activated glycoprotein 1, tax-transcriptionally activated glycoprotein 1 (34kD), tax-transcriptionally activated glycoprotein 1, 34kD, tumor necrosis factor (ligand) superfamily, member 4, tumor necrosis factor ligand superfamily member 4, TXGP1 | |
Mouse myeloma cell line NS0-derived recombinant human OX40 Ligand, Gln51-Leu183, Accession # P23510 | |
0.1 mL | |
Cancer, Cell Cycle and Replication, Immunology | |
7292 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction