Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OX40 Ligand/TNFSF4 Antibody (MM0505-8S23) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP211969
Description
OX40 Ligand/TNFSF4 Monoclonal specifically detects OX40 Ligand/TNFSF4 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Neutralization, CyTOF-ready.Specifications
OX40 Ligand/TNFSF4 | |
Monoclonal | |
LYOPH | |
Western Blot 1:1000, Flow Cytometry 1:50-100, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:20-200, Neutralization 50 - 200 molar excess, CyTOF-ready, Block/Neutralize | |
P23510 | |
TNFSF4 | |
Human recombinant OX40 Ligand/TNFSF4 | |
0.1 mg | |
Primary | |
Please note the larger size(s) come in lyophilized form and the sample size will be in liquid formate. Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml. | |
Store lyophilized antibody at -20°C prior to opening. Aliquot reconstituted liquid and and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Neutralization, CyTOF, Neutralization | |
MM0505-8S23 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS. 0.025 mg size is provided in liquid form, PBS with No Preservative | |
CD134 ligand, CD134L, CD252, CD252 antigen, Glycoprotein Gp34, gp34, OX40 antigen ligand, OX40 ligand, OX40L, OX-40L, OX4OL, TAX transcriptionally-activated glycoprotein 1, tax-transcriptionally activated glycoprotein 1 (34kD), tax-transcriptionally activated glycoprotein 1, 34kD, tumor necrosis factor (ligand) superfamily, member 4, tumor necrosis factor ligand superfamily member 4, TXGP1 | |
Mouse | |
Protein G purified | |
RUO | |
7292 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction