Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OX40 Ligand/TNFSF4 Antibody (MM0505-8S23) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP211969SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211969SS 0.025 mg
NBP211969 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP211969SS Supplier Novus Biologicals Supplier No. NBP211969SS
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

OX40 Ligand/TNFSF4 Monoclonal specifically detects OX40 Ligand/TNFSF4 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Neutralization, CyTOF-ready.

Specifications

Antigen OX40 Ligand/TNFSF4
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Neutralization, CyTOF, Neutralization
Classification Monoclonal
Clone MM0505-8S23
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:1000, Flow Cytometry 1:50-100, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:20-200, Neutralization 50 - 200 molar excess, CyTOF-ready, Block/Neutralize
Formulation Lyophilized from a 0.2 μm filtered solution in PBS. 0.025 mg size is provided in liquid form, PBS with No Preservative
Gene Accession No. P23510
Gene Alias CD134 ligand, CD134L, CD252, CD252 antigen, Glycoprotein Gp34, gp34, OX40 antigen ligand, OX40 ligand, OX40L, OX-40L, OX4OL, TAX transcriptionally-activated glycoprotein 1, tax-transcriptionally activated glycoprotein 1 (34kD), tax-transcriptionally activated glycoprotein 1, 34kD, tumor necrosis factor (ligand) superfamily, member 4, tumor necrosis factor ligand superfamily member 4, TXGP1
Gene Symbols TNFSF4
Host Species Mouse
Immunogen Human recombinant OX40 Ligand/TNFSF4
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7292
Reconstitution Please note the larger size(s) come in lyophilized form and the sample size will be in liquid formate. Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml.
Target Species Human
Content And Storage Store lyophilized antibody at -20°C prior to opening. Aliquot reconstituted liquid and and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.