Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

V alpha 24 J alpha 18 TCR Antibody (6B11) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP200267 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP200267 0.1 mg
NBP200267SS 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP200267 Supplier Novus Biologicals Supplier No. NBP200267
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 12 publications

V alpha 24 J alpha 18 TCR Monoclonal specifically detects V alpha 24 J alpha 18 TCR in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Flow (Cell Surface), CyTOF-ready.

Specifications

Antigen V alpha 24 J alpha 18 TCR
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 6B11
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:10-1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 16867051), Immunohistochemistry-Frozen reported in multiple pieces of scientific literature, Flow (Cell Surface), CyTOF-ready
Formulation PBS with 0.02% Sodium Azide
Gene Alias iNKT, iNKT cell, invariant natural killer T cell, invariant NK T cell, invariant NKT cell
Host Species Mouse
Immunogen Peptide epitope in the CDR3 region of V alpha 24
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.