Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OX40 Ligand/TNFSF4 Antibody (R4930 (Oxelumab)) - Azide and BSA Free, Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals NBP275913SS
Description
OX40 Ligand/TNFSF4 Monoclonal specifically detects OX40 Ligand/TNFSF4 in Human samples. It is validated for Flow Cytometry, Block/Neutralize.Specifications
OX40 Ligand/TNFSF4 | |
Monoclonal | |
Unconjugated | |
PBS with 0.02% Proclin 300 | |
TNFSF4 | |
Oxelumab binds to human OX40L, the ligand of OX40, stimulatory receptor that is expressed on activated T cells, natural killer (NK) cells and natural killer T (NKT) cells. | |
0.025 mg | |
7292.0 | |
Human | |
Purified |
Flow Cytometry, Blocking Assay, Neutralization | |
R4930 (Oxelumab) | |
Flow Cytometry, Block/Neutralize | |
biosimilar, biosimilars, CD134 ligand, CD134L, CD252, CD252 antigen, Glycoprotein Gp34, gp34, OX40 antigen ligand, OX40 ligand, OX40L, OX-40L, OX4OL, TAX transcriptionally-activated glycoprotein 1, tax-transcriptionally activated glycoprotein 1 (34kD), ta | |
Human | |
Protein A purified | |
Primary | |
Binds to human OX40L. | |
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction