Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OX40 Monoclonal Antibody (7D6), FITC
GREENER_CHOICE

Catalog No. PIMA516547
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516547 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516547 Supplier Invitrogen™ Supplier No. MA516547
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody does not react with mouse. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.

OX40 is a protein receptor found on the surface of T cells, a type of white blood cell that plays a crucial role in the immune system. When activated, OX40 promotes the proliferation and survival of T cells, leading to a stronger immune response against cancer cells and infectious agents. OX40 agonists, which are drugs that activate OX40, have shown promising results in preclinical and clinical studies as a potential immunotherapy for cancer. They can enhance the efficacy of other cancer treatments such as chemotherapy and checkpoint inhibitors, and may also have applications in autoimmune diseases and allergies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OX40
Applications Flow Cytometry
Classification Monoclonal
Clone 7D6
Concentration 0.1 mg/mL
Conjugate FITC
Formulation PBS with 1% BSA and 0.09% sodium azide
Gene Tnfrsf4
Gene Accession No. P43489
Gene Alias ACT35; ACT35 antigen; ATC35 antigen; CD134; CD134 antigen; IMD16; Ly-70; lymphoid activation antigene ACT35; MRC OX40; Ox40; OX40 antigen; OX40 cell surface antigen; OX40 homologue; OX40L receptor; TAX transcriptionally-activated glycoprotein 1 receptor; tax-transcriptionally activated glycoprotein 1; tax-transcriptionally activated glycoprotein 1 receptor; TNF receptor superfamily member 4; Tnfrsf4; tumor necrosis factor (ligand) superfamily member 4; tumor necrosis factor receptor superfamily member 4; tumor necrosis factor receptor superfamily, member 4; tumor necrosis factor superfamily, member 4; Txgp1; Txgp1l
Gene Symbols Tnfrsf4
Host Species Mouse
Immunogen CHO-derived feline CD134-Fc fusion protein.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 493665
Target Species Feline, Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.