Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OX40/TNFRSF4 Antibody (7D6), DyLight 650

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10064375C
Description
OX40/TNFRSF4 Monoclonal specifically detects OX40/TNFRSF4 in Feline samples. It is validated for Western Blot, Flow Cytometry.Specifications
OX40/TNFRSF4 | |
Monoclonal | |
DyLight 650 | |
50 mM sodium borate with 0.05% sodium azide | |
TNFRSF4 | |
CHO-derived feline CD134-Fc fusion protein. | |
0.1 mL | |
Primary | |
NB100-64375 recognizes feline CD134, otherwise known as OX40, a 43kDa type I membrane protein and member of the tumor necrosis factor receptor superfamily, expressed predominantly by CD4+ activated T cells, and a key regulator of T cell-dependent immune responses. CD134 has been identified as the receptor for the OX40 ligand, CD252, expressed by activated B cells, acting as a co-stimulatory signal for the stimulation and secretion of immunoglobulins. CD134 has also been identified as a binding receptor for Feline Immunodeficiency Virus (FIV), acting along with CXC chemokine receptor 4 (CXCR4) to facilitate the entry of the virus into CD4+ primary target cells. | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry | |
7D6 | |
Western Blot, Flow Cytometry | |
ACT35 antigen, ACT35ATC35 antigen, CD134, CD134 antigen, OX40 cell surface antigen, OX40 homologue, OX40L receptor, OX40lymphoid activation antigene ACT35, TAX transcriptionally-activated glycoprotein 1 receptor, tax-transcriptionally activated glycoprotein 1 receptor, tumor necrosis factor receptor superfamily member 4, tumor necrosis factor receptor superfamily, member 4, TXGP1LOX40 antigen | |
Mouse | |
Protein A purified | |
Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cell Cycle and Replication, Immunology, Myeloid derived Suppressor Cell, Signal Transduction | |
7293.0 | |
Feline | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction