Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OX40/TNFRSF4 Antibody (977974) [DyLight 594], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB10543M
Description
OX40/TNFRSF4 Monoclonal antibody specifically detects OX40/TNFRSF4 in Human samples. It is validated for Flow Cytometry, Neutralization, CyTOFSpecifications
OX40/TNFRSF4 | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, Neutralization, CyTOF | |
977974 | |
Flow Cytometry, Neutralization, CyTOF-ready | |
ACT35 antigen, ACT35ATC35 antigen, CD134, CD134 antigen, OX40 cell surface antigen, OX40 homologue, OX40L receptor, OX40lymphoid activation antigene ACT35, TAX transcriptionally-activated glycoprotein 1 receptor, tax-transcriptionally activated glycoprotein 1 receptor, tumor necrosis factor receptor superfamily member 4, tumor necrosis factor receptor superfamily, member 4, TXGP1LOX40 antigen | |
Mouse myeloma cell line NS0-derived recombinant human OX40/TNFRSF4, Leu29-Ala216, Accession # P43489 | |
0.1 mL | |
Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cell Cycle and Replication, Immunology, Myeloid derived Suppressor Cell, Signal Transduction | |
7293 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction