Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OX40/TNFRSF4 Antibody (OX-86), DyLight 488, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NB10064847G
Description
OX40/TNFRSF4 Monoclonal specifically detects OX40/TNFRSF4 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
OX40/TNFRSF4 | |
Monoclonal | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
TNFRSF4 | |
CHO cells transfected with murine OX40 | |
0.125 mL | |
Cancer, Cell Cycle and Replication, Immunology | |
7293 | |
Mouse | |
Purified |
Flow Cytometry, Immunohistochemistry (Frozen) | |
OX-86 | |
Flow Cytometry, Immunohistochemistry-Frozen | |
ACT35 antigen, ACT35ATC35 antigen, CD134, CD134 antigen, OX40 cell surface antigen, OX40 homologue, OX40L receptor, OX40lymphoid activation antigene ACT35, TAX transcriptionally-activated glycoprotein 1 receptor, tax-transcriptionally activated glycoprotein 1 receptor, tumor necrosis factor receptor superfamily member 4, tumor necrosis factor receptor superfamily, member 4, TXGP1LOX40 antigen | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
NB100-64847 recognises murine OX40 (CD134), a cell surface antigen expressed only by activated T lymphocytes. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction