Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OX40/TNFRSF4 Antibody (SR1764), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | OX40/TNFRSF4 |
---|---|
Clone | SR1764 |
Dilution | Western Blot 1:500-1:1000, Flow Cytometry 1:50 |
Applications | Western Blot, Flow Cytometry |
Classification | Monoclonal |
Description
OX40/TNFRSF4 Monoclonal antibody specifically detects OX40/TNFRSF4 in Human samples. It is validated for Western Blot, Flow CytometrySpecifications
OX40/TNFRSF4 | |
Western Blot 1:500-1:1000, Flow Cytometry 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
ACT35 antigen, ACT35ATC35 antigen, CD134, CD134 antigen, OX40 cell surface antigen, OX40 homologue, OX40L receptor, OX40lymphoid activation antigene ACT35, TAX transcriptionally-activated glycoprotein 1 receptor, tax-transcriptionally activated glycoprotein 1 receptor, tumor necrosis factor receptor superfamily member 4, tumor necrosis factor receptor superfamily, member 4, TXGP1LOX40 antigen | |
A synthesized peptide derived from human OX40/TNFRSF4 (Uniprot #: P43489) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1764 | |
Western Blot, Flow Cytometry | |
Unconjugated | |
Rabbit | |
Adaptive Immunity, B Cell Development and Differentiation Markers, Cancer, Cell Cycle and Replication, Immunology, Myeloid derived Suppressor Cell, Signal Transduction | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
7293 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title