Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OXA1L Mouse anti-Human, Clone: 4A2D9, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | OXA1L |
---|---|
Clone | 4A2D9 |
Concentration | 0.82 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6564
![]() ![]() |
Proteintech Group Inc
661281IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
OXA1L is required for the insertion of integral membrane proteins into the mitochondrial inner membrane. It is essential for the activity and assembly of cytochrome oxidase and is required for the correct biogenesis of ATP synthase and complex I in mitochondria.Specifications
OXA1L | |
0.82 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Hsa, OXA1 like protein, OXA1Hs, OXA1L | |
OXA1L | |
IgG1 | |
Protein G | |
Antibody |
4A2D9 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q15070 | |
5018 | |
OXA1L Fusion Protein Ag13819 | |
Primary | |
-20°C | |
OXA1L |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title