Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OXA1L Polyclonal Antibody

Catalog No. PIPA542895
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542895 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542895 Supplier Thermo Scientific Supplier No. PA542895

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

OXA1L Polyclonal Antibody

OXA1L Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen OXA1L
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene OXA1L
Gene Accession No. Q15070
Gene Alias 1810020M02Rik; AI894287; Hsa; hypothetical protein LOC520335; LOW QUALITY PROTEIN: mitochondrial inner membrane protein OXA1L; mitochondrial inner membrane protein OXA1L; OXA1; OXA1Hs; Oxa1l; OXA1L mitochondrial inner membrane protein; OXA1L, mitochondrial inner membrane protein; OXA1-like protein; oxidase (cytochrome c) assembly 1-like; oxidase assembly 1-like; oxidase assembly 1-like protein; zgc:163091; zgc:171949
Gene Symbols OXA1L
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human OXA1L (aa 371-420).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5018
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.