Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OXA1L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551953
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551953 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551953 Supplier Invitrogen™ Supplier No. PA551953
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FSLVQVSCLR IPAVRTVLKI PQRVVHDLDK LPPREGFLES FKKGWKNAEM TRQLREREQR MRNQLELAAR GPLRQTFTHN PLLQPGKDNP PNIPSSSSKP KSKYPWHDTL G Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 71%.

OXA1L is required for the insertion of integral membrane proteins into the mitochondrial inner membrane. It is essential for the activity and assembly of cytochrome oxidase and is required for the correct biogenesis of ATP synthase and complex I in mitochondria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OXA1L
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OXA1L
Gene Accession No. Q15070
Gene Alias 1810020M02Rik; AI894287; Hsa; hypothetical protein LOC520335; LOW QUALITY PROTEIN: mitochondrial inner membrane protein OXA1L; mitochondrial inner membrane protein OXA1L; OXA1; OXA1Hs; Oxa1l; OXA1L mitochondrial inner membrane protein; OXA1L, mitochondrial inner membrane protein; OXA1-like protein; oxidase (cytochrome c) assembly 1-like; oxidase assembly 1-like; oxidase assembly 1-like protein; zgc:163091; zgc:171949
Gene Symbols OXA1L
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human OXA1L. Recombinant protein control fragment (Product #RP-89133).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5018
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.