Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxacillin Sodium Salt Monohydrate 98.0+%, TCI America™
SDP

Catalog No. O035325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O035325G 25 g
O03535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O035325G Supplier TCI America Supplier No. O035325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7240-38-2
Molecular Formula C19H23N3NaO6S
Molecular Weight (g/mol) 444.458
MDL Number MFCD00167146
InChI Key VKNIYYBMVNGVMD-MBGFYOAHSA-N
PubChem CID 131673944
IUPAC Name (2S,5R,6R)-3,3-dimethyl-6-[(5-methyl-3-phenyl-1,2-oxazole-4-carbonyl)amino]-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;molecular hydrogen;sodium;hydrate
SMILES [HH].CC1=C(C(=NO1)C2=CC=CC=C2)C(=O)NC3C4N(C3=O)C(C(S4)(C)C)C(=O)O.O.[Na]

Specifications

Melting Point 188°C
Color White-Yellow
Quantity 25 g
Formula Weight 423.42
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Oxacillin Sodium Salt Monohydrate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.