Alfa Aesar™ Oxalacetic acid, 98+%

Manufacturer: Alfa Aesar™ 1578909

Quantity 10g

 View more versions of this product

Catalog No. AA1578909

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 328-42-7
    Molecular Formula C4H4O5
    Formula Weight 132.07
    MDL Number MFCD00002592
    UN Number UN3261
    Beilstein 1705475
    Merck Index 14,6909
    Chemical Name or Material Oxalacetic acid

    Specifications

    Formula Weight 132.07
    Assay Percent Range >98%
    Molecular Formula C4H4O5
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Physical Form Powder
    MDL Number MFCD00002592
    UN Number UN3261
    Beilstein 1705475
    Merck Index 14,6909
    Health Hazard 1 Danger
    Flash Point 88° (190°F)
    Quantity 10g
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    TSCA TSCA
    CAS 328-42-7
    Melting Point 161°C (dec.)
    Chemical Name or Material Oxalacetic acid
    Assay ≥98%

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit