Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxalic acid dihydrate, 99+%, extra pure

Catalog No. p-3586466
Click to view available options
Quantity:
100 g
1 kg
5 kg
Packaging:
Plastic Bottle
Plastic drum

CAS: 6153-56-6 | C2H6O6 | 126.064 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6153-56-6
Molecular Formula C2H6O6
Molecular Weight (g/mol) 126.064
MDL Number MFCD00149102
InChI Key GEVPUGOOGXGPIO-UHFFFAOYSA-N
Synonym oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm
PubChem CID 61373
IUPAC Name oxalic acid;dihydrate
SMILES C(=O)(C(=O)O)O.O.O

Specifications

Melting Point 98.0°C to 102.0°C
Assay Percent Range 99+%
Linear Formula HO2CCO2H·2H2O
Beilstein 02, 502
Fieser 01,764; 02,301; 05,481
Merck Index 15, 7010
Solubility Information Solubility in water: 138g/L (20°C). Other solubilities: 1200g/L in water (90°C), 400g/L in ethanol, 555g/L in boiling alcohol, 181g/L in glycerol, 10g/L in ether
Formula Weight 126.04
Percent Purity ≥99%
Grade Extra Pure
Quantity 100 g
Infrared Spectrum Authentic
Packaging Plastic Bottle
Physical Form Low Melting Crystalline Mass, Powder, Crystals and/or Chunks
Chemical Name or Material Oxalic acid dihydrate
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.