Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Oxalic Acid, Dihydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Shop All Avantor Products
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Specifications
Specifications
Formula Weight | 126.07 |
Grade | BAKER ANALYZED™ ACS Reagent |
Quantity | 500 g |
Packaging | Poly Bottle |
Physical Form | Crystalline |
Specific Gravity | 1L = 1.65kg |
Chemical Name or Material | Oxalic Acid, Dihydrate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction