Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxalic Acid TS, 6.3% (w/v), Ricca Chemical
SDP

Catalog No. p-3252018 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle

Chemical Identifiers

CAS 7732-18-5, 6153-56-6
Molecular Formula C2H6O6
Molecular Weight (g/mol) Mixture
InChI Key GEVPUGOOGXGPIO-UHFFFAOYSA-N
Synonym oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm
PubChem CID 61373
IUPAC Name oxalic acid;dihydrate
SMILES C(=O)(C(=O)O)O.O.O

Specifications

pH 1.3 (0.1M)
Melting Point ∼0°C
Boiling Point ∼100°C
CAS Min % 92.0, 5.81
Color Colorless
CAS Max % 96.0, 6.05
Solubility Information Infinite
Concentration 6.3% (w/v)
Percent Purity 6.3% w/v
Grade Laboratory
Concentration or Composition (by Analyte or Components) 6.3% (w/v)
Quantity 500 mL
Packaging Natural Poly Bottle
Physical Form Liquid
Specific Gravity ∽1.02 to 1.05
Chemical Name or Material Oxalic Acid, Test Solution
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.