Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Oxaloacetic Acid, Calbiochem™,

Catalog No. 50005gm
Click to view available options
Quantity:
5 g

Substrate for malate dehydrogenase and oxaloacetate decarboxylase

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 328-42-7
Molecular Formula C4H4O5
Molecular Weight (g/mol) 132.071
InChI Key KHPXUQMNIQBQEV-UHFFFAOYSA-N
Synonym oxalacetic acid, oxaloacetic acid, 2-oxosuccinic acid, oxaloacetate, ketosuccinic acid, oxosuccinic acid, 2-ketosuccinic acid, butanedioic acid, oxo, oxobutanedioic acid, oxalacetate
PubChem CID 970
ChEBI CHEBI:30744
IUPAC Name 2-oxobutanedioic acid
SMILES C(C(=O)C(=O)O)C(=O)O

Specifications

Melting Point 176°C
Color White
Quantity 5 g
UN Number UN 3261
Solubility Information Easily soluble in cold water
Formula Weight 132.1
Physical Form Solid
Chemical Name or Material Oxaloacetic Acid
DOTInformation DOT Class: 8, Packaging Group: II
Recommended Storage Keep container tightly closed. Keep container in a cool, well-ventilated area. Do not store above 4°C (39.2°F).
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.