Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxamic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
100g
Packaging:
Glass bottle
471-47-6
C2H3NO3
89.05
SOWBFZRMHSNYGE-UHFFFAOYSA-N
2-amino-2-oxoacetic acid,oxamate,acetic acid, aminooxo,oxalamic acid,oxamidic acid,glycine, 2-oxo,oxalic acid monoamide,amino oxo acetic acid,aminooxoacetic acid,oxamate repellent
974
CHEBI:18058
oxamic acid
C(=O)(C(=O)O)N
471-47-6
C2H3NO3
89.05
SOWBFZRMHSNYGE-UHFFFAOYSA-N
2-amino-2-oxoacetic acid,oxamate,acetic acid, aminooxo,oxalamic acid,oxamidic acid,glycine, 2-oxo,oxalic acid monoamide,amino oxo acetic acid,aminooxoacetic acid,oxamate repellent
974
CHEBI:18058
oxamic acid
C(=O)(C(=O)O)N
Melting Point 210.0°C
Packaging Glass bottle
Infrared Spectrum Authentic
Color White
Quantity 10g
Assay Percent Range 98%
Beilstein 02,543
Merck Index 15,7016
Formula Weight 89.05
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material Oxamic acid
Show More Show Less

GHS Signal Word: Warning

EINECSNumber : 207-443-