Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OXCT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543183
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543183 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543183 Supplier Invitrogen™ Supplier No. PA543183
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: GMYANLGIGI PLLASNFISP NITVHLQSEN GVLGLGPYPR QHEADADLIN Sequence homology: Cow: 100%; Dog: 92%; Guinea Pig: 100%; Horse: 92%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 100%; Zebrafish: 92%.

This gene encodes a member of the 3-oxoacid CoA-transferase gene family. The encoded protein is a homodimeric mitochondrial matrix enzyme that plays a central role in extrahepatic ketone body catabolism by catalyzing the reversible transfer of coenzyme A from succinyl-CoA to acetoacetate. Mutations in this gene are associated with succinyl CoA:3-oxoacid CoA transferase deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OXCT1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene OXCT1
Gene Accession No. B2GV06, P55809
Gene Alias 2610008O03Rik; 3-oxoacid CoA transferase 1; 3-oxoacid CoA-transferase 1; 3-oxoacid-CoA transferase 1; EMBL:AAI66478.1}; OXCT; Oxct1; oxct1 {ECO:0000312; Oxct2a; SCOT; SCOT-s; somatic-type succinyl CoA:3-oxoacid CoA-transferase; somatic-type succinyl-CoA:3-oxoacid CoA-transferase; somatic-type succinyl-CoA:3-oxoacid-CoA-transferase; succinyl CoA:3-oxoacid CoA transferase; succinyl-CoA:3-ketoacid CoA transferase; succinyl-CoA:3-ketoacid coenzyme A transferase 1, mitochondrial; succinyl-CoA:3-ketoacid-CoA transferase; succinyl-CoA:3-ketoacid-coenzyme A transferase 1, mitochondrial; Succinyl-CoA:3-ketoacid-coenzyme A transferase 1, mitochondrial-like protein; Unknown (protein for MGC:137519)
Gene Symbols OXCT1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human OXCT1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5019, 690163
Target Species Human, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.