Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OXCT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584985
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584985 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584985 Supplier Invitrogen™ Supplier No. PA584985
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLTILKEEDG DAGKEEDART RIIRR Highest antigen sequence indentity to the following orthologs: Rat - 44%, Mouse - 44%.

Key enzyme for ketone body catabolism. Transfers the CoA moiety from succinate to acetoacetate. Formation of the enzyme-CoA intermediate proceeds via an unstable anhydride species formed between the carboxylate groups of the enzyme and substrate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OXCT2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OXCT2
Gene Accession No. Q9BYC2
Gene Alias 3-oxoacid CoA transferase 2; 3-oxoacid CoA transferase 2A; 3-oxoacid CoA-transferase 2; 3-oxoacid CoA-transferase 2A; 3-oxoacid-CoA transferase 2A; FKSG25; haploid germ cell specific succinyl CoA transferase; Oxct; Oxct2; Oxct2a; Scot; SCOTT; SCOT-t; SCOT-t1; succinyl-CoA:3-ketoacid coenzyme A transferase 2, mitochondrial; Succinyl-CoA:3-ketoacid coenzyme A transferase 2A, mitochondrial; succinyl-CoA:3-ketoacid-coenzyme A transferase 2, mitochondrial; succinyl-CoA:3-ketoacid-coenzyme A transferase 2A, mitochondrial; succinyl-CoA:3-oxoacid CoA-transferase; testis-specific succinyl CoA:3-oxoacid CoA-transferase; testis-specific succinyl CoA:3-oxoacid CoA-transferase 1; testis-specific succinyl-CoA:3-oxoacid CoA-transferase; Testis-specific succinyl-CoA:3-oxoacid CoA-transferase 1
Gene Symbols OXCT2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human OXCT2. Recombinant protein control fragment (Product #RP-108274).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64064
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.