Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OXCT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Rabbit Polyclonal Antibody

Supplier:  Proteintech Group Inc 171221AP150UL

Encompass_Preferred

 View more versions of this product

Catalog No. 50-173-1395


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

Key enzyme for ketone body catabolism. Transfers the CoA moiety from succinate to acetoacetate. Formation of the enzyme-CoA intermediate proceeds via an unstable anhydride species formed between the carboxylate groups of the enzyme and substrate.
Specifications

Specifications

OXCT2
Polyclonal
Unconjugated
OXCT2
3 oxoacid CoA transferase 2, 3 oxoacid CoA transferase 2A, FKSG25, OXCT2, SCOT T
Rabbit
Antigen Affinity Chromatography
RUO
366463, 64059, 64064
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.29 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BYC2, Q9JJN4
OXCT2, Oxct2a
OXCT2 Fusion Protein Ag10810
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.