Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OXER1/5-oxo-ETE GPCR Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297990PCP
Description
OXER1/5-oxo-ETE GPCR Polyclonal antibody specifically detects OXER1/5-oxo-ETE GPCR in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
OXER1/5-oxo-ETE GPCR | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
5-oxo-ETE acid G-protein-coupled receptor 1,5-oxo-ETE G-protein coupled receptor, GPCR, GPR170G-protein coupled receptor R527, G-protein coupled receptor 170, oxoeicosanoid (OXE) receptor 1, oxoeicosanoid receptor 1, TG1019G-protein coupled receptor TG1019 | |
Produced in rabbits immunized with E. coli-derived Human OXER1/5-oxo-ETE GPCR fragment. | |
0.1 mL | |
GPCR | |
165140 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction