Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OXER1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5102296
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5102296 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5102296 Supplier Invitrogen™ Supplier No. PA5102296
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total OXER1.

TG1019 is a G protein-coupled receptor that functions as a chemoattractant for eosinophils and neutrophils. TG1019 is expressed in various tissues except the brain, with the highest expression in liver, kidney, peripheral leukocyte, lung and spleen. TG1019 interacts with the protein OXE to form TG1019/OXE, a Galpha i/o protein-coupled receptor. Signals from TG1019 are transduced via a Galpha i/o protein to PLC/calcium, MEK/ERK and PI3K/Akt pathways. Signal transduction from TG1019 following stimulation with 5-oxo-6E,8Z,11Z,14Z eicosatetraenoic acid (5-oxo-ETE) induces intracellular calcium mobilization and rapid activation of MEK/ERK and PI3K/Akt pathways. TG1019 also may play a role in downregulating cAMP production.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OXER1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene OXER1
Gene Accession No. Q8TDS5
Gene Alias 5-oxo-ETE acid G-protein-coupled receptor 1; 5-oxo-ETE G-protein coupled receptor; GPCR; GPR170; G-protein coupled receptor 170; G-protein coupled receptor R527; G-protein coupled receptor TG1019; HEOAD54; OXE receptor; OXER1; oxoeicosanoid (OXE) receptor 1; oxoeicosanoid receptor 1; TG1019
Gene Symbols OXER1
Host Species Rabbit
Immunogen A synthesized peptide derived from human OXER1(Accession Q8TDS5), corresponding to amino acid residues I220-F270.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 165140
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.