Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OXER1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570736
Description
OXER1 Polyclonal Antibody for Western Blot
TG1019 is a G protein-coupled receptor that functions as a chemoattractant for eosinophils and neutrophils. TG1019 is expressed in various tissues except the brain, with the highest expression in liver, kidney, peripheral leukocyte, lung and spleen. TG1019 interacts with the protein OXE to form TG1019/OXE, a Galpha i/o protein-coupled receptor. Signals from TG1019 are transduced via a Galpha i/o protein to PLC/calcium, MEK/ERK and PI3K/Akt pathways. Signal transduction from TG1019 following stimulation with 5-oxo-6E,8Z,11Z,14Z eicosatetraenoic acid (5-oxo-ETE) induces intracellular calcium mobilization and rapid activation of MEK/ERK and PI3K/Akt pathways. TG1019 also may play a role in downregulating cAMP production.
Specifications
OXER1 | |
Polyclonal | |
Unconjugated | |
OXER1 | |
5-oxo-ETE acid G-protein-coupled receptor 1; 5-oxo-ETE G-protein coupled receptor; GPCR; GPR170; G-protein coupled receptor 170; G-protein coupled receptor R527; G-protein coupled receptor TG1019; HEOAD54; OXE receptor; OXER1; oxoeicosanoid (OXE) receptor 1; oxoeicosanoid receptor 1; TG1019 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
165140 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8TDS5 | |
OXER1 | |
synthetic peptide directed towards the C-terminal region of human OXER1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction