Learn More
Tocris Bioscience™ OXF BD 02
Description
Specifications
Specifications
| Purity | >98% |
| Quantity | 10 mg |
| Product Type | OXF BD 02 |
| Molecular Weight (g/mol) | 295.33 |
| Inhibitors | OXF BD 02 |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.