Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OXGR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532960
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532960 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532960 Supplier Invitrogen™ Supplier No. PA532960
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Marmoset (100%); Monkey, Mouse, Rat, Rabbit (94%); Dog, Bat, Hamster, Panda, Horse (88%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.

G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OXGR1
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene OXGR1
Gene Accession No. Q96P68
Gene Alias 2-oxoglutarate receptor 1; aKGR; Alpha-ketoglutarate receptor 1; Cysltr3; G protein-coupled receptor 80; G protein-coupled receptor 99; Gm299; Gpr80; Gpr99; G-protein coupled receptor 80; G-protein coupled receptor 99; MGC119206; MGC119207; MGC119208; Oxgr1; oxoglutarate (alpha-ketoglutarate) receptor 1; oxoglutarate receptor 1; P2RY15; P2Y purinoceptor 15; P2Y15; P2Y-like GPCR; P2Y-like nucleotide receptor; seven transmembrane helix receptor
Gene Symbols OXGR1
Host Species Rabbit
Immunogen Synthetic 17 amino acid peptide from 2nd extracellular domain of human OXGR1.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27199
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.