Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell Burdick & Jackson™ Oxidation Reagent, 0.05M Iodine in 80/10/10 Acetonitrile/Pyridine/Water (v/v)
Click to view available options
Quantity:
2 x 1 L
4 x 4 L
Packaging Type:
GLASS BOTTLE, GL45
Glass Bottle
Description
Burdick & Jackson™ BioSyn™ reagents & solvents for DNA/RNA synthesis
- Honeywell Burdick & Jackson™ BioSyn™ Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
- ideal for oligonucleotide synthesis regardless of the application
- All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination

Specifications
Specifications
CAS | 75-05-8 |
Content And Storage | Room Temperature |
Density | 0.8159 g/cm3 |
Packaging Type | GLASS BOTTLE, GL45 |
Quantity | 2 x 1 L |
Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction