Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxidation Reagent, Honeywell Burdick & Jackson™

Catalog No. p-7145570 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
4 L
10 L
20 L
200 L
This item is not returnable. View return policy
This item is not returnable. View return policy

BioSyn™, suitable for use with DNA/RNA synthesis instruments, 0.02M Iodine in 70/20/10 THF/Pyridine/Water (v/v)

TRUSTED_SUSTAINABILITY

Specifications

CAS 109-99-9, 110-86-1, 7732-18-5, 7553-56-2
CAS Max % 0.50%, 67.20%, 10.90%, 21.40%
Boiling Point 66°C
Quantity 4 L
Flash Point -15°C
Packaging Returnable Stainless Steel Drum
Physical Form Liquid, clear
Purity Grade Notes Oxidation reagents
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.