Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenoxazine, 97%

Catalog No. AC130220050
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle

CAS: 135-67-1 | C12H9NO | 183.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135-67-1
Molecular Formula C12H9NO
Molecular Weight (g/mol) 183.21
MDL Number MFCD00005014
InChI Key TZMSYXZUNZXBOL-UHFFFAOYSA-N
Synonym phenoxazine, phenazoxine, unii-c2zwt499sg, c2zwt499sg, 5,4-oxazine, phenotazine, rarechem fh 2c 0s92, phenoxazine 1g, 9ci
PubChem CID 67278
IUPAC Name 10H-phenoxazine
SMILES N1C2=CC=CC=C2OC2=CC=CC=C12

Specifications

Melting Point 154.0°C to 159.0°C
Color Gray to Gray-Green
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 27, 62
Merck Index 15, 7365
Solubility Information Solubility in water: insoluble. Other solubilities: freely soluble in abs.methanol, ethanol, ether,, chloroform and benzene, sparingly soluble in petroleum ether
Formula Weight 183.21
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material Phenoxazine
Show More Show Less
EINECSNumber 205-210-8

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.