Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diversey™ Oxivir™ 1 Wipes

Catalog No. p-10501750
Click to view available options
Packaging Quantity:
160 Wipes/Bucket
160 Wipes/Pk., 4 Pk./Cs.
Container Type:
Bag
Bucket

Next generation ready-to-use, pre-wetted, one-step disinfectant cleaner wipe, based on proprietary AHP™ technology. Oxivir™ 1 Wipes are formulated to deliver broad spectrum disinfection in ONE minute or less, with 75 total claims.

  • Multi-surface cleaner/disinfectant wipes
  • Broad spectrum, multi-surface cleaner/disinfectant and sanitizer
  • Harnesses the cleaning power of hydrogen peroxide
  • Specially designed to penetrate the toughest soils
  • Fast, Effective, Responsible and Sustainable formulation
  • Environmentally-responsible with no VOCs, no added fragrance, no NPEs
  • Fast-acting, convenient formula, suitable for daily use, and contact time of 1 minute or less
  • Formulated with patented AHP™ technology containing no phosphates, quats or phenols
  • The active ingredient breaks down to water and oxygen after use

Specifications

Product Type Disinfectant Cleaner Wipes
Color White
Container Type Bucket
For Use With (Application) Cleaning surfaces
Packaging Quantity 160 Wipes/Bucket
Sterility Non-sterile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.