Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OXNAD1 Antibody (OTI1D1), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273175AF700
Description
OXNAD1 Monoclonal specifically detects OXNAD1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
OXNAD1 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
OXNAD1 | |
Full length recombinant protein of human OXNAD1 (NP612390)produced in HEK293 cell. | |
0.1 mL | |
92106.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI1D1 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
MGC15763, oxidoreductase NAD-binding domain containing 1, oxidoreductase NAD-binding domain-containing protein 1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction