Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OXNAD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565180
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565180 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565180 Supplier Invitrogen™ Supplier No. PA565180
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SFKAGQWVDFF IPGVSVVGGF SICSSPRLLE QERVIELAVK YTNHPPALWV HNTCTLDCEV AVRVGGEFFF D Highest antigen sequence identity to the following orthologs - mouse 85%, rat 89%.

OXNAD1 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OXNAD1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene OXNAD1
Gene Accession No. Q96HP4
Gene Alias 2410002F01Rik; AW492292; BC019806; oxidoreductase NAD binding domain containing 1; oxidoreductase NAD-binding domain containing 1; oxidoreductase NAD-binding domain-containing protein 1; Oxnad1
Gene Symbols OXNAD1
Host Species Rabbit
Immunogen Recombinant Human OXNAD1. Recombinant protein control fragment (Product #RP-105928).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92106
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.