Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Oxoid™ Brilliance™ Listeria Differential Supplement
Click to view available options
Quantity:
10/Pk.
Description
Oxoid Brilliance Listeria Differential Supplement is added to Brilliance Listeria Agar Base, Mfr. No. CM1080E and Mfr. No. CM1080B, a chromogenic medium for the isolation and differentiation of Listeria
Listeria monocytogenes is the most common pathogenic Listeria spp. and has been shown to be pathogenic to both man and animals. Some Listeria ivanovii strains also possess lecithinase activity and, although Listeria ivanovii are primarily pathogenic to animals, there are strains which have been shown to cause infection in humans. Brilliance Listeria Agar uses the chromogen X-glucoside for presumptive identification of Listeria spp. This chromogen is cleaved by β-glucosidase which is common to all Listeria species. Other organisms that possess this enzyme, such as enterococci, are inhibited by the selective agents within the medium; lithium chloride, polymyxin B and nalidixic acid, while amphotericin inhibits the growth of any yeasts and moulds present in the sample. Listeria monocytogenes and pathogenic Listeria ivanovii are then further differentiated by their ability to produce the phospholipase enzyme, lecithinase. This enzyme hydrolyses the lecithin in the medium, producing an opaque white halo around the colony. Brilliance Listeria Agar is a modification of the formulation described by Ottaviani and Agosti. As in the original formulation, the medium is designed to identify Listeria spp. based on their utilization of a chromogenic substrate. However, in this modification, the pathogenic Listeria spp. are then further differentiated by the detection of lecithinase (phosphotidylcholine phospholipase C (PCPLC) activity, rather than phosphotidylinositol phospholipase C (PIPLC) activity. Both enzymes, PCPLC and PIPLC, are associated with virulence in Listeria spp., and, therefore, the presence of either enzyme is a useful indicator of pathogenicity.
- Add to Brilliance Listeria Agar Base, Part No. CM1080B
- Each vial supplements 500mL of medium
Specifications
Specifications
| Description | Brilliance Listeria Differential Supplement |
| Form | Liquid |
| Product Type | Microbiology Media |
| Quantity | 10/Pk. |
| Yield | Each vial for 500mL medium |
| Species | Listeria |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction