Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Oxoid™ CCDA Selective Supplement
Click to view available options
Quantity:
10 Vials
10/Pk.
Yield:
Each vial for 500mL medium
For 2L medium
Description
Oxoid CCDA Selective Supplement is used for the isolation of Campylobacter spp., especially C. upsaliensis or Arcobacter.
- For the isolation of Arcobacter spp., add to Arcobacter Broth Base, Part No. CM0965B
- For the isolation of Campylobacter spp., especially C. upsaliensis, add to Campylobacter Blood-Free Selective Agar Base (CCDA Preston), Part No. CM0739B
Specifications
Specifications
| Description | CCDA Selective Supplement |
| Form | Freeze-dried |
| Product Type | Selective Supplement |
| Quantity | 10/Pk. |
| Yield | Each vial for 500mL medium |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction