Learn More
Thermo Scientific™ Oxoid™ Dermasel Selective Supplement
                    Click to view available options
                                Quantity:
                                10/Pk.
                    Description
Oxoid Dermasel Selective Supplement is used for Dermasel Medium, Part No. CM0539B.
Each vial supplements 500mL of medium
            Specifications
Specifications
| Description | Dermasel Selctv Supp. | 
| Form | Freeze-dried | 
| Product Type | Selective Supplement | 
| Quantity | 10/Pk. | 
| Yield | Each vial for 500mL medium | 