Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ IMAGEN™ Parainfluenza Virus Typing Kit using Direct Immunofluorescence Assay

Catalog No. k6104112
Click to view available options
Quantity:
50 Tests
This item is not returnable. View return policy
This item is not returnable. View return policy

Rapidly detect and differentiate Parainfluenza virus types 1, 2, and 3 with this rapid and easy-to-read kit.

Rapidly detect and differentiate Parainfluenza virus types 1, 2 and 3 antigens directly in nasopharyngeal aspirates or in cell culture preparations with Thermo Scientific™ IMAGEN™ Parainfluenza Virus Typing Kit. This direct immunofluorescence assay utilizes type-specific monoclonal antibodies to detect and identify Parainfluenza virus types 1, 2 and 3

Parainfluenza viruses are members of the genus Paramyxovirus classified within the family Paramyxoviridae.1 There are 6 species of Paramyxovirus known to infect humans which include Parainfluenza viruses 1, 2, 3, and 4 (subtypes 4a and 4b), Mumps virus, and Newcastle Disease virus. Of the 4 Parainfluenza viruses, types 1, 2, and 3 are now recognized as a major cause of acute respiratory disease in infants and children.2,3

Use IMAGEN Parainfluenza Kit as a rapid, sensitive, and specific method for detection and differentiation of Parainfluenza virus types 1, 2, and 3 antigens directly in nasopharyngeal aspirates or in cell culture preparations.

  • Rapid results—within a little over 30 minutes of receiving test sample or culture
  • Simple—ready-to-use reagents plus all IMAGEN assays follow the same basic methodology
  • Excellent sensitivity and specificity—high quality, well-characterized antibodies used in the assay provide excellent sensitivity and specificity
  • Easy-to-read results
  • Long shelf life—24 months

The IMAGEN Parainfluenza Kit contains fluorescein isothiocyanate (FITC) conjugated monoclonal antibodies which bind specifically to either Parainfluenza virus types 1, 2 or 3. Specimens are incubated with the FITC conjugated reagents for 15 minutes. Excess reagent is then removed by washing with phosphate-buffered saline (PBS). The stained areas are mounted and viewed using epifluorescence illumination. If Parainfluenza virus types 1, 2, or 3 are present then characteristic apple-green intracellular fluorescence is seen within infected cells, which contrasts with the red background staining of uninfected cells.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description IMAGEN™ Parainfluenza Typing
Detectable Analytes Influenza
Detection Method Nasopharyngeal
Sample Type Nasopharyngeal
Type Reagent
Quantity 50 Tests
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.