Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Oxoid™ Legionella GVPC Selective Supplement
Click to view available options
Quantity:
10/Pk.
Description
Oxoid Legionella GVPC Selective Supplement is used for the isolation of Legionella spp. from environmental samples.
- Add to Legionella Agar Base, Part No. CM1203B
- Each vial supplements 500mL of medium

Specifications
Specifications
Description | Legionella GVPC Selective Supplement |
Form | Freeze-dried |
Product Type | Selective Supplement |
Quantity | 10/Pk. |
Yield | Each vial for 500mL medium |
Species | Legionella |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction